missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilyl)thiophene, 97%

Product Code. 15447678
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15447678

Brand: Thermo Scientific Alfa Aesar H61170.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18245-28-8 | C7H12SSi | 156.318 g/mol

2-(Trimethylsilyl)thiophene is used to produce 2,5-bis-adamantan-1-yl-thiophene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Trimethylsilyl)thiophene is used to produce 2,5-bis-adamantan-1-yl-thiophene.

Notes
Keep container tightly sealed. Store refrigerated conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18245-28-8
Molecular Formula C7H12SSi
Molecular Weight (g/mol) 156.318
MDL Number MFCD00005415
InChI Key OANGLGSZPSFVDY-UHFFFAOYSA-N
Synonym 2-thienyltrimethylsilane, 2-trimethylsilylthiophene, trimethyl thiophen-2-yl silane, silane, trimethyl-2-thienyl, trimethyl-2-thienylsilane, 2-trimethylsilyl thiophene, trimethyl 2-thienyl silane, amtsi021, acmc-1c71q, thien-2-yl-trimethyl-silane
PubChem CID 140360
IUPAC Name trimethyl(thiophen-2-yl)silane
SMILES C[Si](C)(C)C1=CC=CS1

Specifications

Density 0.926
Flash Point 41°C (106°F)
Refractive Index 1.497
Quantity 25 g
UN Number UN1993
Formula Weight 156.32
Percent Purity 97%
Chemical Name or Material 2-(Trimethylsilyl)thiophene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.