Learn More
BINDER™ RS 485 Interface (Two Wire)
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
BINDER Chambers Series MK
Unit Size:
Each
Description
- Additional serial interface can be used parallel to Ethernet
- For Multi Management Software APT-COM™
Specifications
Specifications
| Description | RS 485 interface, 2-wire - Additional serial interface can be used parallel to Ethernet, for Multi Management Software APT-COM™ |
| For Use With (Equipment) | BINDER Chambers Series MK |
| Material | Plastic |
| Type | RS 485 Interface |