Learn More
Ohaus™ 2 x Centrifuge Rack Adapters, Frontier™ R-S16x50/4MI Swing Out Rotors
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ R-S16x50/4MI Swing Out Rotors
Brand:  Ohaus™ 30304375
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15846652
  • 291.00€ / Pack of 2
In Stock
to see stock.
Description
Description
  • Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
  • Suitable for use in Frontier™ R-S16x50/4MI Swing Out Rotors
Specifications
Specifications
FA
29 mm
Rack, 4x50ml D29mm FA, 2/pk
2
4 x 50 mL
R-S16x50/4MI Swing Out Rotors
4