Learn More
Ohaus™ 2 x Centrifuge Racks, Frontier™ R-S4x100/5MIS Swing Out Rotors
Shop All Ohaus Products
Click to view available options
Capacity:
1 x 100 mL
1 x 15 mL
1 x 25 mL
1 x 30 mL
1 x 50 mL (29 mm dia. FA)
1 x 50 mL (29 mm dia. RB)
1 x 50 mL (29 mm dia. Rim)
1 x 50 mL (34 mm dia. RB)
1 x 85 mL
2 x 15 mL (17 mm dia. FA)
2 x 15 mL (17 mm dia. Rim)
3 x 10 mL
4 x 10 mL
4 x 15 mL
4 x 5 to 7 mL
5 x 15 mL
7 x 5 to 7 mL
Unit Size:
Pack of 2
Description
- Allows you to centrifuge tubes of custom size or number
- Suitable for use in buckets for Frontier™ R-S4x100/5MIS Swing Out Rotors
Specifications
Specifications
| Capacity | 1 x 30 mL |
| Max. Tube Diameter | 25 |
| For Use With (Equipment) | R-S4x100/5MIS Swing Out Rotors |
| Description | Rack, 1x30ml D25mm Rim, 2/pk, V2 |
| No. of Places | 1 |
| No. per Pack | 2 |