Learn More
Ohaus™ 2 x Centrifuge Tube Adapters, Frontier™ Fixed Angle Rotors
Shop All Ohaus Products
Click to view available options
Capacity:
1 x 15 mL (17 mm dia. FA)
1 x 15 mL (17 mm dia. RB)
1 x 16 mL
1 x 30 mL
1 x 50 mL (29 mm dia. FA)
1 x 50 mL (29 mm dia. RB)
2 x 15 mL
Unit Size:
Pack of 2
Description
- Accommodates centrifuge tubes that are smaller and/or greater in number than the space provided in the rotor carrier
- Suitable for use in the following Frontier™ rotors:
- R-A4x85/12MI
- R-A6x85/13MI
- R-A6x85/14MI
Specifications
Specifications
| Type | FA |
| Capacity | 1 x 50 mL (29 mm dia. FA) |
| Max. Tube Diameter | 29 mm |
| For Use With (Equipment) | Frontier Fixed-Angle Rotors |
| Description | Adapter, 1x50ml D29mm FA, 2/pk |
| No. of Places | 1 |
| No. per Pack | 2 |