Learn More
Ohaus™ 2 x Centrifuge Tube Adapters, Frontier™ Rotors
Shop All Ohaus Products
Click to view available options
Capacity:
5 mL
7 mL
15 mL
16 mL
30 mL
Unit Size:
Pack of 2
Description
- Accommodates centrifuge tubes that are smaller than the space provided in the rotor carrier.
- Suitable for use in the following Frontier™ rotors:
- R-A12x15/6M
- R-A6x50/6MI
- R-A12x15/6MI
- R-A30x15/4MI
- R-A10x50/10MI
Specifications
Specifications
| Capacity | 5 mL |
| Max. Tube Diameter | 13.5 mm |
| For Use With (Equipment) | Frontier Fixed-Angle Rotors |
| Description | Adapter, D13.5mm, 5ml Tubes x2 |
| No. of Places | 1 |
| No. per Pack | 2 |