Learn More
Labnet™ GyroMini™ Nutating Mixer
Product Code. 11885953
Change view
Click to view available options
Speed Range:
20 rpm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Range unitSize
11885953 20 rpm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 11885953 Supplier Labnet™ Supplier No. S0500230VEU
Only null left
Add to basket
Add to basket
Récupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Ideal for mixing samples in blood and centrifuge tubes. Labnet™ GyroMini™ Nutating Mixer combines the motions of an orbital shaker and a rocker to produce a three-dimensional action.
  • Safe for cold room use
  • Gyratory action provides gentle, thorough mixing without foaming
  • Fixed speed and tilt angle to provide optimal level of agitation
  • Supplied with a dimpled mat for holding tubes firmly in place
Specifications
Rocking Angle 20°
Load Bearing Capacity (Metric) 0.8 kg
Speed Range 20 rpm
Electrical Requirements 230/120 V, 50/60 Hz
Voltage 120/230 V
Temperature Range (Metric) 4°C to 65°C
Frequency 50/60 Hz
Depth (Metric) 171.5 mm
Height (Metric) 152.4 mm
Width (Metric) 241 mm
Includes DSimpled mat