Learn More
Fisherbrand™ Nickel Microspatula
Product Code. 11716447 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
130 mm
Unit Size:
Pack of 5
Product Code. 11716447
Only null left
Add to basket
Add to basket
Use with blances, microscopes and weighing boats. The Fisherbrand™ Nickel Microspatula is designed to dispense and transfer samples during weighing.
  • Dispense and transfer samples during weighing
  • Apply micro amounts of adhesive for tacking down specimens, spreading paste and mixing granular samples
  • Flat-ended blade
Specifications
Quantity 5 Pk.
Product Type Spatula
Finish Bright
For Use With (Application) For Structural Applications Where Resistance to Corrosion is Prime Consideration
Width (Metric) Blade 4 mm
Temperature (Metric) Max 315°C
Material Nickel
Length (Metric) 130 mm