Learn More
Laboratoires Anios 20LT ANIOSYME X3 4X5L
Brand:  Laboratoires Anios 2633036UG
20LT ANIOSYME X3 4X5L
Product Code. 17762688
  • 491.00€ / 20 L
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • ANIOSYME X3
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: ammonio)ethyl]-roxy-poly(oxy-1,2-ethanediyl) propanoate, alcohols, C8-10, e