Learn More
Applied Biosystems™ 21 CFR Part 11 Software Module for QuantStudio™ 6 and 7 Systems
Product Code. 15938506 Shop All Applied Biosystems Products
Click to view available options
Quantity:
10 licenses
Unit Size:
Each
Product Code. 15938506
Brand: Applied Biosystems™ 4486400
This software module for the QuantStudio™ 6 Flex and QuantStudio™ 7 Flex real-time PCR systems supports compliance with 21 CFR Part 11 FDA guidelines on electronic records security and prevents unauthorized access to the instrument.
When loaded on the system, this software will:
• Authenticate each user's log-in information
• Lock out access and require re-authentication if the user is inactive for a set period of time
• Log any unauthorized attempts to access the system software in the Event Log
• Verify each user's permission to perform predefined Controlled Activities
• Permit E-signature function for data verification
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) QuantStudio™ 6 Flex, QuantStudio™ 7
Product Line QuantStudio
Software Type Security
Product Type Software Module
For Use With (Application) Real Time PCR (qPCR)
Quantity 10 licenses