Learn More
Applied Biosystems™ 21 CFR Part 11 Software Module for QuantStudio™ 6 and 7 Systems
Shop All Applied Biosystems Products
Click to view available options
Quantity:
10 licenses
Unit Size:
Each
Description
When loaded on the system, this software will:
• Authenticate each user's log-in information
• Lock out access and require re-authentication if the user is inactive for a set period of time
• Log any unauthorized attempts to access the system software in the Event Log
• Verify each user's permission to perform predefined Controlled Activities
• Permit E-signature function for data verification
Specifications
Specifications
| For Use With (Equipment) | QuantStudio™ 6 Flex, QuantStudio™ 7 |
| Product Line | QuantStudio |
| Software Type | Security |
| Product Type | Software Module |
| For Use With (Application) | Real Time PCR (qPCR) |
| Quantity | 10 licenses |