missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Oxide (Activated, Basic, Brockmann I), TRC
Click to view available options
Quantity:
250 g
Unit Size:
250g
Specifications
Specifications
| Chemical Name or Material | Aluminium oxide |
| CAS | 1344-28-1 |
| Molecular Formula | Al2 O3 |
| Quantity | 250 g |
| Synonym | Aluminium Oxide, Alumina |
| InChI Formula | InChI=1S/2Al.3O |
| SMILES | O=[Al]O[Al]=O |
| IUPAC Name | oxo(oxoalumanyloxy)alumane |
| Molecular Weight (g/mol) | 101.96 |
| Formula Weight | 101.9478 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction