Learn More
1-O-Acetyl-3,5-bis(4-chlorobenzoyl)-2-deoxy-D-ribose, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 1-O-Acetyl-3,5-bis(4-chlorobenzoyl)-2-deoxy-D-ribose |
| CAS | 1207459-15-1 |
| Molecular Formula | C21 H18 Cl2 O7 |
| Quantity | 250 mg |
| Synonym | D-erythro-Pentofuranose, 2-deoxy-, 1-acetate 3,5-bis(4-chlorobenzoate), [(2R,3S)-5-acetoxy-3-(4-chlorobenzoyl)oxy-tetrahydrofuran-2-yl]methyl 4-chlorobenzoate, 1-O-Acetyl-3,5-bis(4-chlorobenzoyl)-2-deoxy-D-ribose |
| InChI Formula | InChI=1S/C21H18Cl2O7/c1-12(24)28-19-10-17(30-21(26)14-4-8-16(23)9-5-14)18(29-19)11-27-20(25)13-2-6-15(22)7-3-13/h2-9,17-19H,10-11H2,1H3/t17-,18+,19?/m0/s1 |
| SMILES | CC(=O)OC1C[C@H](OC(=O)c2ccc(Cl)cc2)[C@@H](COC(=O)c3ccc(Cl)cc3)O1 |
| IUPAC Name | [(2R,3S)-5-acetyloxy-3-(4-chlorobenzoyl)oxyoxolan-2-yl]methyl 4-chlorobenzoate |
| Molecular Weight (g/mol) | 453.27 |
| Formula Weight | 452.043 |