Learn More
Bromopentacarbonylrhenium(I), TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Bromopentacarbonylrhenium(I) |
| CAS | 14220-21-4 |
| Molecular Formula | C5BrO5Re |
| Quantity | 250 mg |
| Synonym | Bromopentacarbonylrhenium, Bromopentacarbonylrhenium(I) |
| InChI Formula | InChI=1S/5CO.BrH.Re/c5*1-2;;/h;;;;;1H;/q;;;;;;+1/p-1 |
| SMILES | [Re]Br.[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+] |
| IUPAC Name | bromorhenium;carbon monoxide |
| Molecular Weight (g/mol) | 406.16 |
| Formula Weight | 405.85 |