Learn More
Butralin, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Butralin |
| CAS | 33629-47-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C14 H21 N3 O4 |
| Quantity | 250 mg |
| Synonym | Aniline, N-sec-butyl-4-tert-butyl-2,6-dinitro- (8CI), 4-(1,1-Dimethylethyl)-N-(1-methylpropyl)-2,6-dinitrobenzenamine, 4-tert-Butyl-N-mono-sec-butyl-2,6-dinitroaniline, 4-tert-Butyl-N-sec-butyl-2,6-dinitroaniline, 70-314B, 72-A34, A 820, A 820 (herbicide), Amchem 70-25, Amex, Amex 820, Butalin, Butralin, Dibutalin, N-sec-Butyl-4-tert-butyl-2,6-dinitroaniline |
| InChI Formula | InChI=1S/C14H21N3O4/c1-6-9(2)15-13-11(16(18)19)7-10(14(3,4)5)8-12(13)17(20)21/h7-9,15H,6H2,1-5H3 |
| SMILES | CCC(C)Nc1c(cc(cc1[N+](=O)[O-])C(C)(C)C)[N+](=O)[O-] |
| IUPAC Name | N-butan-2-yl-4-tert-butyl-2,6-dinitroaniline |
| Molecular Weight (g/mol) | 295.33 |
| Show More |