Learn More
delta-Valerobetaine, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | delta-Valerobetaine |
| CAS | 6778-33-2 |
| Molecular Formula | C8 H17 N O2 |
| Quantity | 250 mg |
| Synonym | delta-Valerobetaine, 5-(trimethylammonio)pentanoate |
| InChI Formula | InChI=1S/C8H17NO2/c1-9(2,3)7-5-4-6-8(10)11/h4-7H2,1-3H3 |
| SMILES | C[N+](C)(C)CCCCC(=O)[O-] |
| IUPAC Name | 5-(trimethylazaniumyl)pentanoate |
| Molecular Weight (g/mol) | 159.226 |
| Formula Weight | 159.126 |