missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N’-Bis(2,4-xylyl)formamidine, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | N,N'-Bis(2,4-Xylyl)Formamidine |
| CAS | 16596-04-6 |
| Molecular Formula | C17 H20 N2 |
| Quantity | 250 mg |
| Synonym | N,N’-Bis(2,4-dimethylphenyl)methanimidamide, N,N'-Di-2,4-xylylformamidine, N,N’-Bis(2,4-dimethylphenyl)formamidine |
| InChI Formula | InChI=1S/C17H20N2/c1-12-5-7-16(14(3)9-12)18-11-19-17-8-6-13(2)10-15(17)4/h5-11H,1-4H3,(H,18,19) |
| SMILES | Cc1ccc(N\C=N\c2ccc(C)cc2C)c(C)c1 |
| IUPAC Name | N,N'-bis(2,4-dimethylphenyl)methanimidamide |
| Molecular Weight (g/mol) | 252.35 |
| Formula Weight | 252.1626 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction