missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone Phenylpropionate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Testosterone Phenylpropionate
Specifications
Specifications
| Chemical Name or Material | Testosterone Phenylpropionate |
| CAS | 1255-49-8 |
| Analyte or Component Names | Testosterone Phenylpropionate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C28 H36 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Testosterone, hydrocinnamate (6CI,7CI,8CI), Hydrocinnamic acid, ester with testosterone (8CI), Androject, Hydrocinnamic acid, testosterone ester, NSC 26643, Retandrol, Testosterone 17-phenylpropionate, Testosterone phenpropionate, Testosterone phenylpropanoate, Testosterone phenylpropionate, Androst-4-en-3-one, 17-(1-oxo-3-phenylpropoxy)-, (17beta)-, (17beta)-17-(1-Oxo-3-phenylpropoxy)androst-4-en-3-one, 3-Oxoandrost-4-en-17beta-yl beta-phenylpropionate |
| InChI Formula | InChI=1S/C28H36O3/c1-27-16-14-21(29)18-20(27)9-10-22-23-11-12-25(28(23,2)17-15-24(22)27)31-26(30)13-8-19-6-4-3-5-7-19/h3-7,18,22-25H,8-17H2,1-2H3/t22-,23-,24-,25-,27-,28-/m0/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2OC(=O)CCc5ccccc5 |
| IUPAC Name | [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] 3-phenylpropanoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction