missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrylamide-d5, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | Acrylamide-d5 |
| CAS | 108152-65-4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C3 D5 N O |
| Quantity | 25 mg |
| Synonym | Acrylamide-d5 (6CI) |
| InChI Formula | InChI=1S/C3H5NO/c1-2-3(4)5/h2H,1H2,(H2,4,5)/i1D2,2D/hD2 |
| SMILES | [2H]N([2H])C(=O)C(=C([2H])[2H])[2H] |
| IUPAC Name | N,N,2,3,3-pentadeuterioprop-2-enamide |
| Molecular Weight (g/mol) | 76.11 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction