missing translation for 'onlineSavingsMsg'
Learn More

Anastrozole Carboxylic Acid (3,5-Bis(1-cyano-1-methylethyl)benzoic Acid), Mikromol™

Product Code. 18001010
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18001010 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18001010 Supplier Mikromol Supplier No. MM1250.010025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Anastrozole Carboxylic Acid (3,5-Bis(1-cyano-1-methylethyl)benzoic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5-Bis(1-cyano-1-methylethyl)benzoic Acid
CAS 1085181-50-5
Analyte or Component Names 3,5-Bis(1-cyano-1-methylethyl)benzoic Acid
Shipping Condition Room Temperature
Molecular Formula C15 H16 N2 O2
Certifications/Compliance ISO 17025
Synonym 3,5-Bis(1-cyano-1-methylethyl)benzoic Acid, 3,5-Bis(2-cyanopropan-2-yl)benzoic Acid
InChI Formula InChI=1S/C15H16N2O2/c1-14(2,8-16)11-5-10(13(18)19)6-12(7-11)15(3,4)9-17/h5-7H,1-4H3,(H,18,19)
SMILES CC(C)(C#N)c1cc(cc(c1)C(C)(C)C#N)C(=O)O
IUPAC Name 3,5-bis(2-cyanopropan-2-yl)benzoic acid
Molecular Weight (g/mol) 256.3
Quantity 25 mg
Formula Weight 256.1212 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.