Learn More
Atomoxetine-d3 Hydrochloride, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | Atomoxetine-d3 Hydrochloride |
| CAS | 1217776-38-9 |
| Molecular Formula | C17H19D3ClNO |
| Quantity | 25 mg |
| Synonym | (R)-N-(Methyl-d3)-(2-methylphenoxy)benzenepropanamine Hydrochloride, R-Tomexetine-d3 Hydrochloride, Tomoxetine-d3 Hydrochloride, LY 139603-d3, Strattera-d3 |
| InChI Formula | InChI=1S/C17H21NO.ClH/c1-14-8-6-7-11-16(14)19-17(12-13-18-2)15-9-4-3-5-10-15;/h3-11,17-18H,12-13H2,1-2H3;1H/t17-;/m1./s1/i2D3; |
| SMILES | CC1=C(O[C@@H](C2=CC=CC=C2)CCNC([2H])([2H])[2H])C=CC=C1.Cl |
| IUPAC Name | (3R)-3-(2-methylphenoxy)-3-phenyl-N-(trideuteriomethyl)propan-1-amine;hydrochloride |
| Molecular Weight (g/mol) | 294.83 |