missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clindamycin Phosphate Sulfoxide trans-N-Oxide, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Clindamycin Phosphate Sulfoxide trans-N-Oxide
Specifications
Specifications
| Chemical Name or Material | Clindamycin Phosphate Sulfoxide trans-N-Oxide |
| Analyte or Component Names | Clindamycin Phosphate Sulfoxide trans-N-Oxide |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C18 H34 Cl N2 O10 P S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Clindamycin Phosphate Sulfoxide trans-N-Oxide |
| InChI Formula | InChI=1S/C18H34ClN2O10PS/c1-5-6-10-7-11(21(3,25)8-10)17(24)20-12(9(2)19)15-13(22)14(23)16(31-32(26,27)28)18(30-15)33(4)29/h9-16,18,22-23H,5-8H2,1-4H3,(H,20,24)(H2,26,27,28)/t9-,10+,11-,12+,13+,14-,15+,16+,18+,21-,33?/m0/s1 |
| SMILES | CCC[C@@H]1C[C@@H](C(=O)N[C@H]([C@H](C)Cl)[C@H]2O[C@@H]([C@H](OP(=O)(O)O)[C@@H](O)[C@H]2O)S(=O)C)[N@@+](C)([O-])C1 |
| IUPAC Name | [(2R,3R,4S,5R,6R)-6-[(1S,2S)-2-chloro-1-[[(1S,2S,4R)-1-methyl-1-oxido-4-propylpyrrolidin-1-ium-2-carbonyl]amino]propyl]-4,5-dihydroxy-2-methylsulfinyloxan-3-yl] dihydrogen phosphate |
| Molecular Weight (g/mol) | 536.96 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction