missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxybupropion (as (RS,RS)-cyclic Hemiketal), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Hydroxybupropion (as (RS,RS)-cyclic Hemiketal)
Specifications
Specifications
| Chemical Name or Material | Hydroxybupropion (as (RS,RS)-cyclic Hemiketal) |
| CAS | 233600-52-7 |
| Analyte or Component Names | Hydroxybupropion (as (RS,RS)-cyclic Hemiketal) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H18 Cl N O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Hydroxybupropion (as (RS,RS)-cyclic Hemiketal) |
| InChI Formula | InChI=1S/C13H18ClNO2/c1-9-13(16,17-8-12(2,3)15-9)10-5-4-6-11(14)7-10/h4-7,9,15-16H,8H2,1-3H3/t9-,13+/m0/s1 |
| SMILES | C[C@@H]1NC(C)(C)CO[C@@]1(O)c2cccc(Cl)c2 |
| IUPAC Name | (2S,3S)-2-(3-chlorophenyl)-3,5,5-trimethylmorpholin-2-ol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction