missing translation for 'onlineSavingsMsg'
Learn More

Liothyronine Methyl Ester, Mikromol™

Product Code. 30052207
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30052207

Brand: Mikromol MM0626.090025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Liothyronine Methyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Liothyronine Methyl Ester
CAS 54914-53-3
Analyte or Component Names Liothyronine Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C16 H14 I3 N O4
Certifications/Compliance ISO 9001
Synonym Liothyronine Methyl Ester, O-(4-hydroxy-3-iodophenyl)-3,5-diiodo-L-Tyrosine methyl ester, 3,5,3'-Triiodo-L-thyronine methyl ester, L-Triiodothyronine methyl ester
InChI Formula InChI=1S/C16H14I3NO4/c1-23-16(22)13(20)6-8-4-11(18)15(12(19)5-8)24-9-2-3-14(21)10(17)7-9/h2-5,7,13,21H,6,20H2,1H3/t13-/m0/s1
SMILES COC(=O)[C@@H](N)Cc1cc(I)c(Oc2ccc(O)c(I)c2)c(I)c1
IUPAC Name methyl (2S)-2-amino-3-[4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl]propanoate
Molecular Weight (g/mol) 665
Quantity 25 mg
Formula Weight 664.8057 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.