Learn More
Code Corporation™ 2700 Handled Barcode Scanner
Product Code. p-200020005
Click to view available options
Includes:
B27 Battery
B27 Battery, Inductive Charging Station, 1.8 m USB Charging Cable
Unit Size:
Each
Regulatory: This product is under eco-contribution tax.
Product Code. 17963862
Brand: Code Corporation™ 176687
Regulatory: This product is under eco-contribution tax.
Easily read wide and small barcodes with the ultra-fast CR2700 Handled Barcode Scanner. This aggressive, omnidirectional scanner eliminates repositioning and time-consuming re-scans of barcodes.
  • Easily read wide and small barcodes with the ultra-fast CR2700 Handled Barcode Scanner.
  • This aggressive, omnidirectional scanner eliminates repositioning and time-consuming re-scans of barcodes.
  • Get your scanning processes started or add this scanner + battery kit to your current scanning solution.
  • Uses B27 batteries with built-in power gauge.
  • IP65 seal rating against dust and liquid ingress, CodeShield™ housing for disinfectant and chemical wipedown.
  • Unequaled performance reading on curved and shiny surfaces.
  • USB, Bluetooth™5 Low Energy (for enhanced data security, extended battery life and less downtime).
  • Customisable visual, audible and haptic feedback with multiple programmable buttons.
  • Trace any troubles – Trackable service dates and battery statistics to help IT deploy and manage their scanner fleet.
Specifications
Description Code Reader™ 2700 Handled Barcode Scanner with Charging Station
Supported Barcode Types BC412, Codabar, Code 11, Code 128, Code 32, Code 39, Code 93, GS1 DataBar, Hong Kong 2 of 5, IATA 2 of 5, Interleaved 2 of 5, MSI Plessey, Matrix 2 of 5, NEC 2 of 5, Pharmacode, Plessey, Straight 2 of 5, Telepen, Trioptic, UPC / EAN / JANCodablock F, Code 49, GS1 Composite (CC-A/CC-B/CC-C), MicroPDF, PDF417Aztec Code, Data Matrix, Data Matrix Rectangular Ext'n, GoCode&trade, (prorietary--add'l license req'd), Grid Matrix, HanXin Code, Micro QR Code, QR Code, QR Model 1Australian Post, Canada Post, Itelligent Mail, Japan Post, KIX Code, Korea Post, Planet, Post-Net, UK Royal Mail, UPU ID-tags
Data Transfer Method Bluetooth 5 Low Energy (Class II)
Height (Metric) 135.00 mm
Height (English) 5.3 in.
Width (Metric) 53.34 mm
Width (English) 2.1 in.
Depth (Metric) 131.00 mm
Depth (English) 5.2 in.
Weight (Metric) 177 g
Charging Connection USB 2.0 (Generic HID, HID Keyboard, Virtual COM Port)
Weight (English) 0.29 lb.
Certifications/Compliance CE, Europe: CE-RoHS
For Use With (Application) Asset Tracking, General Identification Healthcare, Laboratory, Manufacturing, Pharmaceutical
Humidity 5% to 95% (Non-condensing)
Includes B27 Battery, Inductive Charging Station, 1.8 m USB Charging Cable
Required Accessories Bluetooth 5 Low Energy (Class II)
Warranty 3 Years
Software Type JavaScript
Quantity 1 Kit
Type Barcode Scanner
Connection Type Bluetooth
Display Type None Included
Power Supply Rechargeable Battery
Alternate Part Number CR2702-100-A271-C36-MB6-C286