Learn More
3M™ 2840 Series Polycarbonate Lens Safety Glasses
Product Code. 10237824
Click to view available options
Lens Tint:
Amber
Clear
Mirror
Unit Size:
Each
Pack of 20
Product Code. 10237824
Brand: 3M™ 7000032459
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Specifications
Certifications/Compliance EN 166:2001
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Safety glasses; Safety Spectacles
For Use With (Application) Construction,‎ pharmaceuticals
Frame Color Silver
No. per Pack 20/Pk.
Temple Style Adjustable length
Lens Material Polycarbonate