missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2E)-3-(Dimethylamino)-1-(pyridin-3-yl)prop-2-en-1-one, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
(2E)-3-(Dimethylamino)-1-(pyridin-3-yl)prop-2-en-1-one
Specifications
Specifications
| Chemical Name or Material | (2E)-3-(dimethylamino)-1-(pyridin-3-yl)prop-2-ene-1-one |
| CAS | 123367-26-0 |
| Analyte or Component Names | (2E)-3-(dimethylamino)-1-(pyridin-3-yl)prop-2-ene-1-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H12 N2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Imatinib Mesilate Imp. A (EP), (2E)-3-(Dimethylamino)-1-(pyridin-3-yl)prop-2-ene-1-one, 3-Dimethylamino-1-(3-pyridyl)-2-propen-1-one, Imatinib Imp. A (EP), 2-Propen-1-one, 3-(dimethylamino)-1-(3-pyridinyl)-, (2E)-, Imatinib Mesilate Impurity A, Imatinib Impurity A |
| InChI Formula | InChI=1S/C10H12N2O/c1-12(2)7-5-10(13)9-4-3-6-11-8-9/h3-8H,1-2H3/b7-5+ |
| SMILES | CN(C)\C=C\C(=O)c1cccnc1 |
| IUPAC Name | (E)-3-(dimethylamino)-1-pyridin-3-ylprop-2-en-1-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction