missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S-Venlafaxine-di-p-toluoyl-L-tartrate Salt (2:1), TRC
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Specifications
Specifications
| Chemical Name or Material | S-Venlafaxine-di-p-toluoyl-L-tartrate Salt |
| CAS | 93471-25-1 |
| Molecular Formula | C54H72N2O12 |
| Quantity | 2 mg |
| Synonym | 1-[(1S)-2-(Dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexanol (2R,3R)-2,3-Bis[(4-methylbenzoyl)oxy]butanedioate Salt |
| InChI Formula | InChI=1S/C20H18O8.2C17H27NO2/c1-11-3-7-13(8-4-11)19(25)27-15(17(21)22)16(18(23)24)28-20(26)14-9-5-12(2)6-10-14;2*1-18(2)13-16(17(19)11-5-4-6-12-17)14-7-9-15(20-3)10-8-14/h3-10,15-16H,1-2H3,(H,21,22)(H,23,24);2*7-10,16,19H,4-6,11-13H2,1-3H3/t15-,16-;2*16-/m111/s1 |
| SMILES | OC1(CCCCC1)[C@H](CN(C)C)C2=CC=C(OC)C=C2.OC([C@H](OC(C3=CC=C(C)C=C3)=O)[C@@H](OC(C4=CC=C(C)C=C4)=O)C(O)=O)=O.OC5(CCCCC5)[C@H](CN(C)C)C6=CC=C(OC)C=C6 |
| IUPAC Name | (S)-1-(2-(dimethylamino)-1-(4-methoxyphenyl)ethyl)cyclohexan-1-ol hemi((2R,3R)-2,3-bis((4-methylbenzoyl)oxy)succinate) |
| Molecular Weight (g/mol) | 941.16 |
| Formula Weight | 940.51 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction