missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2RS)-2-Cyclopentyl-2-phenylacetic Acid, Mikromol™
Description
(2RS)-2-Cyclopentyl-2-phenylacetic Acid
Specifications
Specifications
| Chemical Name or Material | (2RS)-2-Cyclopentyl-2-phenylacetic Acid |
| CAS | 3900-93-4 |
| Analyte or Component Names | (2RS)-2-Cyclopentyl-2-phenylacetic Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H16 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (2RS)-2-Cyclopentyl-2-phenylacetic Acid, Glycopyrronium Bromide Imp. K (EP), Glycopyrronium Bromide Impurity K, Glycopyrronium Impurity K |
| InChI Formula | InChI=1S/C13H16O2/c14-13(15)12(11-8-4-5-9-11)10-6-2-1-3-7-10/h1-3,6-7,11-12H,4-5,8-9H2,(H,14,15) |
| SMILES | OC(=O)C(C1CCCC1)c2ccccc2 |
| IUPAC Name | 2-cyclopentyl-2-phenylacetic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?