missing translation for 'onlineSavingsMsg'
Learn More

Melford 2XYT, Capsule

Product Code. 30018928
Click to view available options
Quantity:
496 G
992 g
Unit Size:
496g
992g
This item is not returnable. View return policy

Product Code. 30018928

Brand: Melford X156401000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Nutritionally enriched medium for the growth of recombinant strains of E.

Nutritionally enriched medium for the growth of recombinant strains of E. Coli and in the preparation and transformation of competent cells.

Material is specially granulated to reduce dust and increase solubility rate.

Dissolve 31g in one liter distilled water. Heat solution to boiling with continuous stirring. Sterilize by autoclaving at 121°C for 15 minutes. Final pH 25°C: 6.8-7.2.

Ingredients:

Digest Peptone: 16 g/l

Yeast Extract: 10 g/l

Sodium Chloride: 5 g/l

Specifications

Quantity 992 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.