missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(1H-1,2,4-Triazol-1-yl)benzoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | 3-1h-1,2,4-triazol-1-yl benzoic acid, 3-1,2,4-triazol-1-yl benzoic acid, 3-1,2,4 triazol-1-yl-benzoic acid, benzoic acid, 3-1h-1,2,4-triazol-1-yl, pubchem19567, 3-1,2,4-triazolyl benzoic acid, 1-3-carboxyphenyl-1h-1,2,4-triazole, benzoic acid,3-1h-1,2,4-triazol-1-yl |
Formula Weight | 189.17 |
Percent Purity | 97% |
Chemical Name or Material | 3-(1H-1, 2, 4-Triazol-1-yl)benzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'