missing translation for 'onlineSavingsMsg'
Learn More

[3-(1H-1,2,4-Triazol-1-yl)phenyl]methanol, ≥95%, Thermo Scientific™

Catalog Number p-3176176
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
868755-55-9
C9H9N3O
175.19
MFCD08271897
KUSZGGXHHOYFHS-UHFFFAOYSA-N
3-1h-1,2,4-triazol-1-yl phenyl methanol, 3-1,2,4-triazol-1-yl phenyl methanol, 3-1,2,4-triazolyl phenyl methan-1-ol
20485344
[3-(1H-1,2,4-triazol-1-yl)phenyl]methanol
OCC1=CC(=CC=C1)N1C=NC=N1
This item is not returnable. View return policy
868755-55-9
C9H9N3O
175.19
MFCD08271897
KUSZGGXHHOYFHS-UHFFFAOYSA-N
3-1h-1,2,4-triazol-1-yl phenyl methanol, 3-1,2,4-triazol-1-yl phenyl methanol, 3-1,2,4-triazolyl phenyl methan-1-ol
20485344
[3-(1H-1,2,4-triazol-1-yl)phenyl]methanol
OCC1=CC(=CC=C1)N1C=NC=N1
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 175.19
Percent Purity 95%
Chemical Name or Material [3-(1H-1, 2, 4-triazol-1-yl)phenyl]methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'