missing translation for 'onlineSavingsMsg'
Learn More

3-(1H-Imidazol-1-ylmethyl)benzoic acid hydrochloride, Tech., Thermo Scientific™

Catalog Number p-3175362
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
218131-32-9
C11H11ClN2O2
238.671
MFCD08690248
YTCNDYWVEAFCMT-UHFFFAOYSA-N
3-1h-imidazol-1-ylmethyl benzoic acid hydrochloride, 3-imidazol-1-ylmethyl-benzoic acid hydrochloride, 3-imidazol-1-ylmethyl benzoic acid hydrochloride, 3-imidazolylmethyl benzoic acid, chloride, 3-1h-imidazol-1-yl methyl benzoic acid hydrochloride, 3-1h-imidazol-1-yl methyl benzoic acid-hydrogen chloride 1/1
18525763
3-(imidazol-1-ylmethyl)benzoic acid;hydrochloride
C1=CC(=CC(=C1)CN2C=CN=C2)C(=O)O.Cl
This item is not returnable. View return policy
218131-32-9
C11H11ClN2O2
238.671
MFCD08690248
YTCNDYWVEAFCMT-UHFFFAOYSA-N
3-1h-imidazol-1-ylmethyl benzoic acid hydrochloride, 3-imidazol-1-ylmethyl-benzoic acid hydrochloride, 3-imidazol-1-ylmethyl benzoic acid hydrochloride, 3-imidazolylmethyl benzoic acid, chloride, 3-1h-imidazol-1-yl methyl benzoic acid hydrochloride, 3-1h-imidazol-1-yl methyl benzoic acid-hydrogen chloride 1/1
18525763
3-(imidazol-1-ylmethyl)benzoic acid;hydrochloride
C1=CC(=CC(=C1)CN2C=CN=C2)C(=O)O.Cl
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Quantity 250 mg
Formula Weight 238.67
Percent Purity 90%
Grade Technical
Chemical Name or Material 3-(1H-Imidazol-1-ylmethyl)benzoic acid hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'