missing translation for 'onlineSavingsMsg'
Learn More

3-(2,2,2-Trifluoroethoxy)benzeneboronic acid, 97%

Product Code. 15499897
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15499897

Brand: Thermo Scientific Alfa Aesar H53137.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 850593-08-7 | C8H8BF3O3 | 219.954 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850593-08-7
Molecular Formula C8H8BF3O3
Molecular Weight (g/mol) 219.954
MDL Number MFCD07363846
InChI Key SZLRKQKOUYNFDB-UHFFFAOYSA-N
Synonym 3-2,2,2-trifluoroethoxy phenylboronic acid, 3-2,2,2-trifluoroethoxy phenyl boronic acid, 3-trifluoroethoxy phenylboronic acid, 3-2,2,2-trifluoroethoxy benzeneboronic acid, boronic acid,b-3-2,2,2-trifluoroethoxy phenyl, 1-2,2,2-trifluoro-ethoxy-benzene-3-boronic acid, acmc-209q3j, 3-trifluoroethoxyphenylboronic acid, 3-2,2,2-trifluoroethoxyl phenylboronic acid
PubChem CID 43145319
IUPAC Name [3-(2,2,2-trifluoroethoxy)phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)OCC(F)(F)F)(O)O

Specifications

Melting Point 126°C to 129°C
Quantity 1 g
Formula Weight 219.95
Percent Purity 97%
Chemical Name or Material 3-(2,2,2-Trifluoroethoxy)benzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.