missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[2-[4-(6-Fluoro-1,2-benzoxazol-3-yl)-1-piperidyl]ethyl]-9-hydroxy-2-methyl-pyrido[1,2-a]pyrimidin-4-one, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
3-[2-[4-(6-Fluoro-1,2-benzoxazol-3-yl)-1-piperidyl]ethyl]-9-hydroxy-2-methyl-pyrido[1,2-a]pyrimidin-4-one
Specifications
Specifications
| Chemical Name or Material | 3-[2-[4-(6-Fluoro-1,2-benzoxazol-3-yl)-1-piperidyl]ethyl]-9-hydroxy-2-methyl-pyrido[1,2-a]pyrimidin-4-one |
| CAS | 766485-15-8 |
| Analyte or Component Names | 3-[2-[4-(6-Fluoro-1,2-benzoxazol-3-yl)-1-piperidyl]ethyl]-9-hydroxy-2-methyl-pyrido[1,2-a]pyrimidin-4-one |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H23 F N4 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4H-Pyrido[1,2-a]pyrimidin-4-one, 3-[2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-9-hydroxy-2-methyl-, 3-[2-[4-(6-Fluoro-1,2-benzoxazol-3-yl)-1-piperidyl]ethyl]-9-hydroxy-2-methyl-pyrido[1,2-a]pyrimidin-4-one, 3-[2-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-9-hydroxy-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one |
| InChI Formula | InChI=1S/C23H23FN4O3/c1-14-17(23(30)28-9-2-3-19(29)22(28)25-14)8-12-27-10-6-15(7-11-27)21-18-5-4-16(24)13-20(18)31-26-21/h2-5,9,13,15,29H,6-8,10-12H2,1H3 |
| SMILES | CC1=C(CCN2CCC(CC2)c3noc4cc(F)ccc34)C(=O)N5C=CC=C(O)C5=N1 |
| IUPAC Name | 3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)-1-piperidyl]ethyl]-9-hydroxy-2-methyl-pyrido[1,2-a]pyrimidin-4-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction