missing translation for 'onlineSavingsMsg'
Learn More

3-(2,4,6-Trimethoxyphenyl)propionic acid, 96%, Thermo Scientific™

Catalog Number 15407147
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
74737-05-6
C12H16O5
240.255
MFCD00045944
LKDSCRCVTKEXRX-UHFFFAOYSA-N
3-2,4,6-trimethoxyphenyl propanoic acid, 3-2,4,6-trimethoxyphenyl propionic acid, 2,4,6-trimethoxyhydrocinnamic acid, 3-2,4,6-trimethoxy-phenyl-propionic acid
3018538
3-(2,4,6-trimethoxyphenyl)propanoic acid
COC1=CC(=C(C(=C1)OC)CCC(=O)O)OC
This item is not returnable. View return policy
74737-05-6
C12H16O5
240.255
MFCD00045944
LKDSCRCVTKEXRX-UHFFFAOYSA-N
3-2,4,6-trimethoxyphenyl propanoic acid, 3-2,4,6-trimethoxyphenyl propionic acid, 2,4,6-trimethoxyhydrocinnamic acid, 3-2,4,6-trimethoxy-phenyl-propionic acid
3018538
3-(2,4,6-trimethoxyphenyl)propanoic acid
COC1=CC(=C(C(=C1)OC)CCC(=O)O)OC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 140°C to 144°C
Quantity 5 g
Formula Weight 240.26
Percent Purity 96%
Chemical Name or Material 3-(2,4,6-Trimethoxyphenyl)propionic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'