Learn More
Alfa Aesar™ 3-(2-Bromophenyl)-1-propanol, 98%
Brand: Alfa Aesar™ H63884.03
Additional Details : CAS : 52221-92-8 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
52221-92-8 | |
215.09 | |
VODAJGPTULSNSU-UHFFFAOYSA-N | |
10512846 | |
C1=CC=C(C(=C1)CCCO)Br |
C9H11BrO | |
MFCD09028685 | |
3-2-bromo-phenyl-propan-1-ol, 3-2-bromophenyl propan-1-ol, 3-2-bromophenyl-1-propanol, benzenepropanol, 2-bromo, ksc268e5r, 3-2-bromo-phenyl ;-propan-1-ol | |
3-(2-bromophenyl)propan-1-ol |
Specifications
3-(2-Bromophenyl)-1-propanol | |
1g | |
C9H11BrO | |
3-2-bromo-phenyl-propan-1-ol, 3-2-bromophenyl propan-1-ol, 3-2-bromophenyl-1-propanol, benzenepropanol, 2-bromo, ksc268e5r, 3-2-bromo-phenyl ;-propan-1-ol | |
C1=CC=C(C(=C1)CCCO)Br | |
215.09 | |
215.09 | |
98% |
Colorless | |
52221-92-8 | |
MFCD09028685 | |
VODAJGPTULSNSU-UHFFFAOYSA-N | |
3-(2-bromophenyl)propan-1-ol | |
10512846 | |
Liquid |