Learn More
3-(2-Methyl-4-pyrimidinyl)benzenesulfonyl chloride, Technical, Thermo Scientific™
Product Code. 10582173
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. 10582173
Brand: Thermo Scientific Maybridge CC31603DA
Request Bulk or Custom Format
Chemical Identifiers
CAS 465514-07-2
Molecular Formula C11H9ClN2O2S
Molecular Weight (g/mol) 268.715
InChI Key ZLLPUDLQNCNHFM-UHFFFAOYSA-N
Synonym 3-2-methylpyrimidin-4-yl benzene-1-sulfonylchloride, 3-2-methyl-4-pyrimidinyl benzenesulfonyl chloride, 3-2-methylpyrimidin-4-yl benzenesulfonyl chloride, 3-2-methylpyrimidin-4-yl benzene-1-sulfonyl chloride, 3-2-methylpyrimidin-4-yl benzenesulphonyl chloride, chloro 3-2-methylpyrimidin-4-yl phenyl sulfone, 3-2-methyl-4-pyrimidinyl phenylsulfonyl chloride, benzenesulfonylchloride, 3-2-methyl-4-pyrimidinyl, benzenesulfonyl chloride, 3-2-methyl-4-pyrimidinyl
PubChem CID 2779830
IUPAC Name 3-(2-methylpyrimidin-4-yl)benzenesulfonyl chloride
SMILES CC1=NC=CC(=N1)C2=CC(=CC=C2)S(=O)(=O)Cl
Specifications
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 268.72
Percent Purity 95%
Chemical Name or Material 3-(2-Methyl-4-pyrimidinyl)benzenesulfonyl chloride