missing translation for 'onlineSavingsMsg'
Learn More

3-(2-Trimethylsiloxyphenyl)-1-propylboronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number p-7030555
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
2096331-55-2
C18H31BO3Si
334.34
MFCD12546185
WVIXWRBQCZVWHF-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-3-2-trimethylsiloxyphenyl propyl-1,3,2-dioxaborolane, 3-2-trimethylsiloxyphenyl-1-propylboronic acid pinacol ester
73995112
This item is not returnable. View return policy
2096331-55-2
C18H31BO3Si
334.34
MFCD12546185
WVIXWRBQCZVWHF-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-3-2-trimethylsiloxyphenyl propyl-1,3,2-dioxaborolane, 3-2-trimethylsiloxyphenyl-1-propylboronic acid pinacol ester
73995112

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Refractive Index 1.4799
Quantity 1 g
Sensitivity Moisture sensitive
Formula Weight 334.34
Percent Purity 95%
Chemical Name or Material 3-(2-Trimethylsiloxyphenyl)-1-propylboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'