missing translation for 'onlineSavingsMsg'
Learn More

(3,3,3-Trifluoropropyl)methyldichlorosilane, 97%

Product Code. p-7046876
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11399305

Brand: Thermo Scientific Alfa Aesar L16669.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 675-62-7 | C4H7Cl2F3Si | 211.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 675-62-7
Molecular Formula C4H7Cl2F3Si
Molecular Weight (g/mol) 211.08
MDL Number MFCD00039294
InChI Key OHABWQNEJUUFAV-UHFFFAOYSA-N
Synonym 3,3,3-trifluoropropyl methyldichlorosilane, 3,3,3-trifluoropropyl dichloromethylsilane, dichloro methyl 3,3,3-trifluoropropyl silane, unii-38xgw8m24k, silane, dichloromethyl 3,3,3-trifluoropropyl, dichloromethyl-3,3,3-trifluoropropylsilane, dichloromethyl 3,3,3-trifluoropropyl silane, dichloro-methyl-3,3,3-trifluoropropyl silane, 4-04-00-04169 beilstein handbook reference, ksc491g7h
PubChem CID 12666
IUPAC Name dichloro-methyl-(3,3,3-trifluoropropyl)silane
SMILES C[Si](CCC(F)(F)F)(Cl)Cl

Specifications

Boiling Point 121°C to 122°C
Quantity 5 g
UN Number UN2985
Beilstein 1744964
Flash Point 15°C (59°F)
Formula Weight 211.09
Percent Purity 97%
Refractive Index 1.385
Sensitivity Moisture sensitive
Density 1.261
Chemical Name or Material (3,3,3-Trifluoropropyl)methyldichlorosilane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.