missing translation for 'onlineSavingsMsg'
Learn More

(3,3,3-Trifluoropropyl)methyldichlorosilane, TRC

Product Code. 30452998
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30452998

Brand: Toronto Research Chemicals TRCT7920055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3,3,3-Trifluoropropyl)methyldichlorosilane
CAS 675-62-7
Molecular Formula C4H7Cl2F3Si
Quantity 5 g
Synonym Dichloromethyl(3,3,3-trifluoropropyl)silane, Methyl-3,3,3-trifluoropropyldichlorosilane
InChI Formula InChI=1S/C4H7Cl2F3Si/c1-10(5,6)3-2-4(7,8)9/h2-3H2,1H3
SMILES C[Si](Cl)(Cl)CCC(F)(F)F
IUPAC Name dichloro-methyl-(3,3,3-trifluoropropyl)silane
Molecular Weight (g/mol) 211.09
Formula Weight 209.9646
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.