Learn More
Lenz Laborglasinstrumente™ 3.3 Borosilicate Glass 1-Way Vacuum Stopcocks
Click to view available options
Plug Bore:
2.5 mm
4 mm
Includes:
Right-angled Sidearm, Hollow Glass Plug
Straight Sidearms, Hollow Glass Plug
Unit Size:
Each
Specifications
Specifications
| Type | 1-way Vacuum Stopcock |
| Plug Bore | 2.5 mm |
| Material | Borosilicate Glass |
| Certifications/Compliance | DIN 12545 |
| Includes | Right-angled Sidearm, Hollow Glass Plug |