Learn More
Lenz Laborglasinstrumente™ 3.3 Borosilicate Glass 3-Way Stopcocks
Click to view available options
Plug Bore:
2.5 mm
4 mm
Includes:
PTFE Plug, Screw Thread Retaining Device
Solid Glass Plug, Screw Thread Retaining Device
Solid Glass Plug, Screw Thread Rubber Retaining Device
Unit Size:
Each
Specifications
Specifications
| Type | 3-way Stopcock |
| Plug Bore | 2.5 mm |
| Material | Borosilicate Glass |
| Certifications/Compliance | DIN 12554 |
| Includes | PTFE Plug, Screw Thread Retaining Device |