Learn More
Alfa Aesar™ 3,3'-Diethyloxacarbocyanine iodide, 98%
Brand: Alfa Aesar™ H55607.03
Additional Details : CAS : 905-96-4 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
905-96-4 | |
461.323 | |
FIZZUEJIOKEFFZ-UHFFFAOYSA-N | |
129893627 | |
CCN1C2=CC=CC=C2OC1=CC=CC3=[N+](C4=CC=CC=C4O3)CC.I |
C21H22IN2O2+ | |
MFCD00011952 | |
3,3'-diethyloxacarbocyanine iodide | |
(2Z)-3-ethyl-2-[(E)-3-(3-ethyl-1,3-benzoxazol-3-ium-2-yl)prop-2-enylidene]-1,3-benzoxazole;hydroiodide |
Specifications
3,3'-Diethyloxacarbocyanine iodide | |
Red | |
1g | |
C21H22IN2O2+ | |
3,3'-diethyloxacarbocyanine iodide | |
CCN1C2=CC=CC=C2OC1=CC=CC3=[N+](C4=CC=CC=C4O3)CC.I | |
461.323 | |
460.31 | |
98% |
Light sensitive | |
278°C (decomposition) | |
905-96-4 | |
MFCD00011952 | |
FIZZUEJIOKEFFZ-UHFFFAOYSA-N | |
(2Z)-3-ethyl-2-[(E)-3-(3-ethyl-1,3-benzoxazol-3-ium-2-yl)prop-2-enylidene]-1,3-benzoxazole;hydroiodide | |
129893627 | |
Powder |