missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(3-Methylphenyl)propionic acid, 96%

Brand:  Alfa Aesar™ H34395.06

Additional Details : CAS : 3751-48-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15407337

  • 171.00€ / 5g


Estimated Shipment
26-04-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

3751-48-2
164.204
VWXVTHDQAOAENP-UHFFFAOYSA-N
138042
CC1=CC(=CC=C1)CCC(=O)O
C10H12O2
MFCD00060326
3-3-methylphenyl propionic acid, 3-3-methylphenyl propanoic acid, 3-m-tolyl propanoic acid, 3-m-tolylpropionic acid, 3-m-tolylpropanoic acid, propionic acid, 3-3-methylphenyl, 3-m-tolyl-propionic acid, 3-methylhydrocinnamic acid, hydrocinnamicacid,m-methyl, 3-methylbenzenepropanoic acid
3-(3-methylphenyl)propanoic acid

Specifications

3-(3-Methylphenyl)propionic acid
5g
C10H12O2
3-3-methylphenyl propionic acid, 3-3-methylphenyl propanoic acid, 3-m-tolyl propanoic acid, 3-m-tolylpropionic acid, 3-m-tolylpropanoic acid, propionic acid, 3-3-methylphenyl, 3-m-tolyl-propionic acid, 3-methylhydrocinnamic acid, hydrocinnamicacid,m-methyl, 3-methylbenzenepropanoic acid
CC1=CC(=CC=C1)CCC(=O)O
164.204
164.2
42°C to 44°C
3751-48-2
MFCD00060326
VWXVTHDQAOAENP-UHFFFAOYSA-N
3-(3-methylphenyl)propanoic acid
138042
96%
Videos
SDS Documents