Learn More
3,3'-Thiodipropionic acid, 98%, Thermo Scientific Chemicals
Catalog Number p-7023550
18.80 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
250 g
1000 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
111-17-1
C6H10O4S
178.202
MFCD00002781
ODJQKYXPKWQWNK-UHFFFAOYSA-N
3,3'-thiodipropionic acid, thiodipropionic acid, propanoic acid, 3,3'-thiobis, bis 2-carboxyethyl sulfide, tyox a, thiodihydracrylic acid, 4-thiaheptanedioic acid, tdpa, sulfide, bis 2-carboxyethyl, 3,3'-thiodipropanoic acid
8096
3-(2-carboxyethylsulfanyl)propanoic acid
C(CSCCC(=O)O)C(=O)O
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
111-17-1
C6H10O4S
178.202
MFCD00002781
ODJQKYXPKWQWNK-UHFFFAOYSA-N
3,3'-thiodipropionic acid, thiodipropionic acid, propanoic acid, 3,3'-thiobis, bis 2-carboxyethyl sulfide, tyox a, thiodihydracrylic acid, 4-thiaheptanedioic acid, tdpa, sulfide, bis 2-carboxyethyl, 3,3'-thiodipropanoic acid
8096
3-(2-carboxyethylsulfanyl)propanoic acid
C(CSCCC(=O)O)C(=O)O
3,3'-Thiodipropionic acid is used as an antioxidant for fats and other foodstuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,3′-Thiodipropionic acid is used as an antioxidant for fats and other foodstuffs.
Solubility
Soluble in acetone, ethanol. Slightly soluble in water.
Notes
Stable under ordinary conditions. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Melting Point 127°C to 134°C
Linear Formula S(CH2CH2CO2H)2
Quantity 250 g
Beilstein 1210299
Merck Index 14,9332
Solubility Information Soluble in acetone,ethanol. Slightly soluble in water.
Formula Weight 178.21
Percent Purity 98%
Chemical Name or Material 3,3'-Thiodipropionic acid