missing translation for 'onlineSavingsMsg'
Learn More

3',4',5,7-Tetrahydroxyflavone, 97%, Thermo Scientific Chemicals

Catalog Number p-7058412
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
491-70-3
C15H10O6
286.239
MFCD00017309
IQPNAANSBPBGFQ-UHFFFAOYSA-N
luteolin, 3',4',5,7-tetrahydroxyflavone, 2-3,4-dihydroxyphenyl-5,7-dihydroxy-4h-chromen-4-one, digitoflavone, luteolol, flacitran, luteoline, weld lake, cyanidenon 1470, salifazide
5280445
CHEBI:15864
2-(3,4-dihydroxyphenyl)-5,7-dihydroxychromen-4-one
C1=CC(=C(C=C1C2=CC(=O)C3=C(C=C(C=C3O2)O)O)O)O
This item is not returnable. View return policy
491-70-3
C15H10O6
286.239
MFCD00017309
IQPNAANSBPBGFQ-UHFFFAOYSA-N
luteolin, 3',4',5,7-tetrahydroxyflavone, 2-3,4-dihydroxyphenyl-5,7-dihydroxy-4h-chromen-4-one, digitoflavone, luteolol, flacitran, luteoline, weld lake, cyanidenon 1470, salifazide
5280445
CHEBI:15864
2-(3,4-dihydroxyphenyl)-5,7-dihydroxychromen-4-one
C1=CC(=C(C=C1C2=CC(=O)C3=C(C=C(C=C3O2)O)O)O)O

3',4',5,7-Tetrahydroxyflavone is used as an apoptosis inducer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′,4′,5,7-Tetrahydroxyflavone is used as an apoptosis inducer.

Solubility
Soluble in aqueous alkaline solutions (1.4 mg/ml), ethanol (∼5 mg/ml), dimethyl sulfoxide (7 mg/ml), 1eq. Sodium hydroxide (5 mM), dimethylformamide (∼20 mg/ml), water (1 mg/ml) at 25°C and methanol.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C.
TRUSTED_SUSTAINABILITY
Melting Point >300°C
Quantity 100 mg
Beilstein 292084
Merck Index 14,5614
Solubility Information Soluble in aqueous alkaline solutions (1.4mg/ml),ethanol (∽5mg/ml),dimethyl sulfoxide (7mg/ml),1eq. Sodium hydroxide (5 mM),dimethylformamide (∽20mg/ml),water (1mg/ml) at 25°C and methanol.
Formula Weight 286.24
Percent Purity 97%
Chemical Name or Material 3',4',5,7-Tetrahydroxyflavone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'