missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluorobenzaldehyde, 98%, Thermo Scientific™

Catalog Number p-7025465
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
132123-54-7
C7H3F3O
160.10
MFCD00083523
NLAVHUUABUFSIG-UHFFFAOYSA-N
benzaldehyde, 3,4,5-trifluoro, 3,4,5-trifluoro-benzaldehyde, 3,4,5-ttrifluorobenzaldehyde, acmc-209bon, intermediates-zcf02014, 3,4,5-triflourobenzaldehyde, 3,4,5-trifluorobrnzaldehyde, ksc182i0f, 3,4,5-trifluorobenzaldehyde, benzaldehyde, 3,4,5-trifluoro-9ci
2776954
3,4,5-trifluorobenzaldehyde
FC1=CC(C=O)=CC(F)=C1F
This item is not returnable. View return policy
132123-54-7
C7H3F3O
160.10
MFCD00083523
NLAVHUUABUFSIG-UHFFFAOYSA-N
benzaldehyde, 3,4,5-trifluoro, 3,4,5-trifluoro-benzaldehyde, 3,4,5-ttrifluorobenzaldehyde, acmc-209bon, intermediates-zcf02014, 3,4,5-triflourobenzaldehyde, 3,4,5-trifluorobrnzaldehyde, ksc182i0f, 3,4,5-trifluorobenzaldehyde, benzaldehyde, 3,4,5-trifluoro-9ci
2776954
3,4,5-trifluorobenzaldehyde
FC1=CC(C=O)=CC(F)=C1F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 24°C to 26°C
Density 1.42
Boiling Point 174°C
Flash Point 62°C (144°F)
Refractive Index 1.478
Quantity 1 g
Beilstein 7805841
Sensitivity Air Sensitive
Formula Weight 160.1
Percent Purity 98%
Chemical Name or Material 3,4,5-Trifluorobenzaldehyde
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'