missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Bromophenyl)-5-methyl-4-phenyl-4H-1,2,4-triazole, 97%, Thermo Scientific™

Catalog Number 15405666
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
1858241-81-2
C15H12BrN3
314.186
MFCD22683118
TYXCTRHZBUPEHB-UHFFFAOYSA-N
3-4-bromophenyl-5-methyl-4-phenyl-4h-1,2,4-triazole, 3-4-bromophenyl-5-methyl-4-phenyl-1,2,4-triazole
98000572
3-(4-bromophenyl)-5-methyl-4-phenyl-1,2,4-triazole
CC1=NN=C(N1C2=CC=CC=C2)C3=CC=C(C=C3)Br
This item is not returnable. View return policy
1858241-81-2
C15H12BrN3
314.186
MFCD22683118
TYXCTRHZBUPEHB-UHFFFAOYSA-N
3-4-bromophenyl-5-methyl-4-phenyl-4h-1,2,4-triazole, 3-4-bromophenyl-5-methyl-4-phenyl-1,2,4-triazole
98000572
3-(4-bromophenyl)-5-methyl-4-phenyl-1,2,4-triazole
CC1=NN=C(N1C2=CC=CC=C2)C3=CC=C(C=C3)Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 100 mg
Formula Weight 314.18
Percent Purity 97%
Chemical Name or Material 3-(4-Bromophenyl)-5-methyl-4-phenyl-4H-1,2,4-triazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'