missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichloro-5-hydroxy-2(5H)-furanone, Mikromol™

Product Code. 17920486
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 17920486

Brand: Mikromol MM3451.110025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

3,4-Dichloro-5-hydroxy-2(5H)-furanone
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,4-Dichloro-5-hydroxy-2(5H)-furanone
CAS 766-40-5
Analyte or Component Names 3,4-Dichloro-5-hydroxy-2(5H)-furanone
Shipping Condition Room Temperature
Molecular Formula C4 H2 Cl2 O3
Certifications/Compliance ISO 9001
Synonym 2(5H)-Furanone, 3,4-dichloro-5-hydroxy-, Crotonic acid, 2,3-dichloro-4,4-dihydroxy-, γ-lactone (7CI,8CI), 3,4-Dichloro-5-hydroxy-2(5H)-furanone, 5-Hydroxy-3,4-dichloro-2-(5H)-furanone, Mucochloric acid lactone, NSC 179787
InChI Formula InChI=1S/C4H2Cl2O3/c5-1-2(6)4(8)9-3(1)7/h3,7H
SMILES OC1OC(=O)C(=C1Cl)Cl
IUPAC Name 3,4-dichloro-2-hydroxy-2H-furan-5-one
Molecular Weight (g/mol) 168.9629
Quantity 25 mg
Formula Weight 167.9381 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.