Learn More
3,4-Dichlorobenzyl alcohol, 98%, Thermo Scientific™
Product Code. 11440763
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. Quantity unitSize
11440763 10 g 10g
11450763 50 g 50g
11460763 250 g 250g
3 options
Product Code. 11440763
Brand: Thermo Scientific Alfa Aesar A13236.09
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1805-32-9
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.024
MDL Number MFCD00004633
InChI Key FVJIUQSKXOYFKG-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzyl alcohol, 3,4-dichlorophenyl methanol, benzenemethanol, 3,4-dichloro, 3,4-dichlorobenzylalcohol, rarechem al bd 0536, benzyl alcohol, 3,4-dichloro, labotest-bb lt02003790, benzenemethanol,4-dichloro, benzyl alcohol,4-dichloro, 3,4-dichloro-1-hydroxymethyl-benzene
PubChem CID 15728
IUPAC Name (3,4-dichlorophenyl)methanol
SMILES C1=CC(=C(C=C1CO)Cl)Cl
Specifications
Melting Point 36°C to 39°C
Boiling Point 130°C (3 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
UN Number UN2811
Beilstein 2082362
Formula Weight 177.03
Percent Purity 98%
Chemical Name or Material 3,4-Dichlorobenzyl alcohol